Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

105° Vacuum Take-Off Adapters, Chemglass

Supplier: Chemglass

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CG-1050-70 CG-1050-71 CG-1050-45 CG-1050-72 CG-1050-19 CG-1050-01 CG-1050-02 CG-1050-14
80066-058EA 112.85 USD InStock
80066-058 16002-650 80066-056 80066-054 89028-244 80066-060 89028-248 89028-246
105° Vacuum Take-Off Adapters, Chemglass
Distillation Components Distillation Adapters
Adapters are bent at a 105° angle for use in vacuum distillation.

  • 105° angle bend
  • Top outer [ST] joint
  • Bottom inner [ST] joint
  • Sealed-in drip tip
  • Serrated hose connection

Adapters feature [ST] top outer and lower inner joints with sealed-in drip tip. Serrated hose connection has a 10 mm O.D. at the largest point of contact to vacuum source.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty