Would you like to log in again?
Vacuum Filtration Adapters, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1053-17 CG-1053-10 CG-1053-12
80066-100EA 179.28 USD InStock
80066-100 80066-086 80066-090
Vacuum Filtration Adapters, Chemglass
Adapters Adapters, Joints with Tube Connectors
Adapters are designed for use with reduced-pressure filtration with plain stem buchner funnels.
  • Flared top
  • [ST] joint
  • With hose connection
Top of adapter is flared to accept the listed pluro stopper size. The hose connection has a 10mm O.D. at the largest serration.
Ordering information: Adapters are not supplied with rubber pluro stoppers.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty