Would you like to log in again?
Inert Gas Inner Joint Adapters, Chemglass
Supplier: Chemglass
CG-1064-01 CG-1064-02 CG-1064-19 CG-1064-14 CG-1064-45
16002-652EA 146.92 USD InStock
16002-652 80066-130 80066-140 80066-128 80066-138
Inert Gas Inner Joint Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters have two hose connections 180° from one another to permit a flow of inert gas over the apparatus.
Hose connection has an O.D. of 10 mm at the largest serration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty