Would you like to log in again?
Distillation Adapters, 105° Bend, with Drip Tips, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1071-A-14 CG-1071-A-19
80066-166EA 107.68 USD InStock
80066-166 80066-164
Distillation Adapters, 105° Bend, with Drip Tips, Chemglass
Distillation Components Distillation Adapters
Distillation adapters with sealed-in lower drip tip and top outer joints, both of same size.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty