Would you like to log in again?
Inlet Adapter, Bleed Tube, Chemglass
Supplier: Chemglass
CG-1078-01
80066-194EA 12.08 USD InStock
80066-194
Inlet Adapter, Bleed Tube, Chemglass
Adapters Adapters, Conical Ground Joints
Use this inlet adapter for the introduction of gases or liquids during reactions.
  • Tapered delivery end
  • Capillary tip
Bleed tube has an O.D. of 7mm with a tapered delivery end and capillary tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty