Would you like to log in again?
Beakers, Jacketed, Conical Bottom, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1103-03 CG-1103-04 CG-1103-05 CG-1103-02 CG-1103-06 CG-1103-01
80066-224EA 637.81 USD InStock
80066-224 80066-222 80066-220 80066-218 80066-216 80066-226
Beakers, Jacketed, Conical Bottom, Chemglass
Beakers
Designed for use in temperature controlled chemical and biological reactions.
Bottom interior surface is conical in shape to help eliminate spinout when used with egg shaped spinbars. Hose connections have an O.D. of 10 mm at the largest serration.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty