Would you like to log in again?
Drechel Gas Washing Bottles, Tall Form, Chemglass
Supplier: Chemglass
CG-1112-04 CG-1112-01 CG-1112-05 CG-1112-02 CG-1112-03 CG-1112-08 CG-1112-07 CG-1112-09 CG-1112-06
80066-234EA 213.73 USD InStock
80066-234 80066-244 80066-232 80066-242 80066-230 80066-240 80066-228 80066-238 80066-236
Drechel Gas Washing Bottles, Tall Form, Chemglass
Bottles Gas Washing Bottles
Tall form gas washing bottle having a standard taper 34/28 top joint, hex base, and plain tube for gas dispersion.
Stopper is held in place with two 18 mm stainless steel springs.
Note: springs are only supplied when bottle is ordered complete.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty