Would you like to log in again?
Gas Washing Bottle, Fritted, Chemglass
Supplier: Chemglass
CG-1120-01 CG-1120-02
80066-352EA 940.76 USD InStock
80066-352 80066-350
Gas Washing Bottle, Fritted, Chemglass
Bottles Gas Washing Bottles
Large capacity, low-form gas washing bottle has a 34/28 standard taper top outer joint.
Filter candle is 25 Ext.Ø×70 mm long, allowing a large gas flow to be dispersed throughout the absorbing material.
Ordering information: Complete bottle is supplied with one each size 34 Keck clamp. Available with a coarse or extra coarse porosity filter candle.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty