Would you like to log in again?
Hybridization Bottles, Chemglass
Supplier: Chemglass
CG-1140-13 CG-1140-12 CG-1140-11 CG-1140-14 CG-1140-03 CG-1140-04 CG-1140-07 CG-1140-16 CG-1140-05 CG-1140-17 CG-1140-01 CG-1140-02 CG-1140-08 CG-1140-10
80066-418EA 202.79 USD InStock
80066-418 80066-428 80066-416 80066-426 80066-414 80066-436 80066-424 80066-434 80066-412 80066-422 80066-432 80066-410 80066-420 80066-430
Hybridization Bottles, Chemglass
Bottles Hybridization Bottles/Tubes
Heavy-wall bottles are used in hybridization incubators or ovens with rotators.
  • Bottle fits most hybridization equipment
  • Straight interior walls for easy membrane placement and removal
  • Dual chamber bottles have a capped opening at each end with a solid glass wall in the center to create two equal compartments
  • Red cap has a PTFE liner and working temperature of 180 °C
  • Blue cap has silicone sealing ring and a working temperature of 140 °C
An alternative to plastic bags or boxes, the membrane is rolled and placed into the bottle which is secured in a rotisserie. The membrane is saturated and drained during each revolution, resulting in even exposure of the blots to probe or wash solutions. Bottle fits most hybridization equipment and has straight interior walls for easy membrane placement and removal, as well as decanting of hybridization solutions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty