Would you like to log in again?
Adapter, Flow Control, Standard Taper Joints, Chemglass
Supplier: Chemglass
CG-1191-02
80066-750EA 263.77 USD InStock
80066-750
Adapter, Flow Control, Standard Taper Joints, Chemglass
Adapters Adapters, Spherical Ground Joints
Flow control adapter for use with CG-1188 and CG-1189 chromatography columns along with CG-1190 reservoirs.
  • Adapter has a 0-4 mm Chem-Cap® metering plug
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty