Would you like to log in again?
Chromatography Columns, with Rodaviss® Joint, PTFE Stopcock and Fritted Disc
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1193-25 CG-1193-16 CG-1193-12 CG-1193-04 CG-1193-07 CG-1193-15 CG-1193-24 CG-1193-19 CG-1193-31 CG-1193-13 CG-1193-05 CG-1193-01 CG-1193-03 CG-1193-10 CG-1193-11 CG-1193-08 CG-1193-30 CG-1193-26 CG-1193-29 CG-1193-20 CG-1193-21
80066-818EA 308.53 USD InStock
80066-818 80066-828 80066-838 80066-826 80066-836 80066-814 80066-834 80066-844 80066-822 80066-832 80066-854 80066-820 80066-842 80066-864 80066-874 80066-852 80066-862 80066-850 80066-872 80066-860 80066-870
Chromatography Columns, with Rodaviss® Joint, PTFE Stopcock and Fritted Disc
Chromatography Columns
General purpose column has a PTFE stopcock and a top Rodaviss® outer joint, with the addition of a sealed-in coarse porosity fritted disc to support column packing.
  • Column is constructed using medium wall tubing
  • Lower end of the column has a round bottom
Can be used as a gravity eluted column or a flash column, when used with optional CG-1195 flow controller and CG-1194 reservoir.
Connections between the column, reservoir and flow control adapter are made using Rodaviss® joints. The unique design of the Rodaviss® joint affords a grease free seal while holding glassware together without clamps. Column is supplied complete with a CG-182 open top cap and O-ring.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty