Would you like to log in again?
Flow Control Adapters with Spherical Joints, Chemglass
Supplier: Chemglass
CG-1201-10 CG-1201-09 CG-1201-11
80067-112EA 246.69 USD InStock
80067-112 80067-114 80067-116
Flow Control Adapters with Spherical Joints, Chemglass
Adapters Adapters, Spherical Ground Joints
These flow control adapters are for use with Chemglass absorption chromatography columns.
  • Chem-Cap® metering plug
  • Replacement plugs available
Adapters have a Chem-Cap® metering plug. Replacement plugs are available.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty