Would you like to log in again?
Condensers, Friedrichs, Spiral Inner Tube, Chemglass
Supplier: Chemglass
CG-1210-02
80067-266EA 565.82 USD InStock
80067-266
Condensers, Friedrichs, Spiral Inner Tube, Chemglass
Condensers
Condensers feature a spiral inner tube.
  • Condenser has a standard taper outer joint at a 15° angle to the condenser
  • Lower inner drip tip joint
Providing a long vapor path, ensuring good heat transfer and anti-flooding characteristics. Hose connections have an O.D. of 10 mm at the middle serration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty