Would you like to log in again?
Distilling Bulbs, Kugelrohr, Chemglass
Supplier: Chemglass
CG-1227-11 CG-1227-12 CG-1227-10 CG-1227-13
80067-474EA 98.56 USD InStock
80067-474 80067-476 80067-470 80067-472
Distilling Bulbs, Kugelrohr, Chemglass
Distillation Components Distillation Receivers
Kugelrohr bulbs feature tapered joints.
These Kugelrohr bulbs having 14/20 standard taper joints on both ends is to connect to other bulbs or single neck round bottom flasks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty