Would you like to log in again?
Distilling Columns, Hemple, Chemglass
Supplier: Chemglass
CG-1228-09 CG-1228-08 CG-1228-05 CG-1228-11
80067-486EA 143.36 USD InStock
80067-486 80067-480 80067-482 80067-478
Distilling Columns, Hemple, Chemglass
Distillation Components Distillation Columns
Distillations feature a row of indentations.
  • Plain distillation column
  • Includes a row of indentations to support column packing material
Column length listed is for the straight portion above the row of indentations and below the joint. Column has a standard taper top outer and lower inner joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty