Would you like to log in again?
Short Path Distilling Heads, Chemglass
Supplier: Chemglass
CG-1239-02 CG-1239-12 CG-1239-11 CG-1239-03 CG-1240-11
80067-584EA 519.49 USD InStock
80067-584 80067-572 80067-574 80067-582 80067-614
Short Path Distilling Heads, Chemglass
Distillation Components Distillation Heads
Designed for use with small quantities where hold-up between the flask and receiver must be kept to a minimum
The thermometer bulb and stem serve as column packing. Upper hose connections are for cooling lines while the lower connection is for vacuum. Available with Vigreux indentations to improve efficiency. Also available vacuum-jacketed to further improve efficiency.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty