Would you like to log in again?
Receiver, Moisture Test, Barrett-Type, Chemglass
Supplier: Chemglass
CG-1261-21 CG-1261-22 CG-1261-25 CG-1261-20 CG-1261-12 CG-1261-07 CG-1261-27 CG-1261-26 CG-1261-11
80067-760EA 393.09 USD InStock
80067-760 80067-762 80067-768 80067-778 80067-758 80067-764 80067-774 80067-766 80067-776
Receiver, Moisture Test, Barrett-Type, Chemglass
Distillation Components Distillation Receivers
This moisture test receiver is used to determine the water content in petroleum and bituminous material.
  • Available with 2mm glass or PTFE stopcock
Receiver is made according to ASTM specifications, with the addition of the top outer standard taper joint and lower stopcock. The 2mL has sub-divisions of 0.02mL; the 5mL has subdivisions of 0.20mL; 10mL has subdivisions of 0.1mL; the 20mL has subdivisions of 0.2 from 0 to 3mL and 0.5 from 3 to 20mL.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty