Would you like to log in again?
Distilling Receivers, Moisture Test, Merlic, Chemglass
Supplier: Chemglass
CG-1264-04 CG-1264-06
80067-826EA 489.66 USD InStock
80067-826 80067-830
Distilling Receivers, Moisture Test, Merlic, Chemglass
Distillation Components Distillation Receivers
Distillation receivers feature efficient cooling.
  • Graduated from 0 to 10 ml in 0.2 ml subdivisions
Modified Dean-Stark trap allows for more efficient cooling and separation of the azeotrope during distillation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty