Would you like to log in again?
Distillation Receiver, Solvent Recovery, Graduated, Chemglass
Supplier: Chemglass
CG-1272-02
80067-858EA 1062.89 USD InStock
80067-858
Distillation Receiver, Solvent Recovery, Graduated, Chemglass
Distillation Components Distillation Receivers
Graduated distillation receiver feature a 4 mm three way stopcock.
  • Upper sampling port has a 2 mm PTFE stopcock
  • The top outer and lower inner joints are the same size
Three way stopcock to permit solvent removal through the 10 mm O.Ø. tube or return to distillation flask.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty