Would you like to log in again?
Snyder Distillation Columns, Improved, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1304-01
80068-010EA 210.21 USD InStock
80068-010
Snyder Distillation Columns, Improved, Chemglass
Distillation Components Distillation Columns
Distillation columns featuring tree section snyder column.
  • Improved design to prevent column flooding
Column has a 24/40 top outer joint and a 24/40 lower inner joint for use with Kuderna-Danish flask.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty