Would you like to log in again?
Filter Apparatus, Chemglass
Supplier: Chemglass
CG-1404-03 CG-1404-02 CG-1404-01
80068-332EA 265.8 USD InStock
80068-332 80068-336 80068-334
Filter Apparatus, Chemglass
Funnels Filtration Funnels Filter Funnels
Useful for direct filtration applications of small volumes under vacuum.
  • Complete apparatus consists of a Buchner funnel
  • Featuring 14/20 top outer and lower inner joints
Erlenmeyer flask 50 ml having a 14/20 outer joint.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty