Would you like to log in again?
Filter Funnels, Pressure, Chemglass
Supplier: Chemglass
CG-1405-16 CG-1405-14 CG-1405-01 CG-1405-07 CG-1405-10 CG-1405-12 CG-1405-03 CG-1405-05
80068-350EA 1185.5 USD InStock
80068-350 80068-352 80068-358 80068-346 80068-356 80068-344 10170-824 80068-348
Filter Funnels, Pressure, Chemglass
Funnels Filtration Funnels Filter Funnels
For positive pressure filtration of materials up to 15 psi.
  • The 24/40 lower inner standard taper joint has an outlet for connection to a vacuum source
  • Complete item consists of a filter funnel, top inlet adapter, Viton O-ring and a stainless steel pinch clamp
Connection between the funnel and adapter is made by an O-ring joint which permits a larger, grease free opening for sample retrieval.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty