Would you like to log in again?
Buchner Funnels, Inner Joint, Chemglass
Supplier: Chemglass
CG-1406-50 CG-1406-44 CG-1406-02 CG-1406-34 CG-1406-55 CG-1406-10 CG-1406-30 CG-1406-21 CG-1406-33 CG-1406-25 CG-1406-45 CG-1406-58 CG-1406-65 CG-1406-49 CG-1406-41 CG-1406-43 CG-1406-04 CG-1406-40 CG-1406-42 CG-1406-07 CG-1406-48 CG-1406-32 CG-1406-47 CG-1406-46 CG-1406-12 CG-1406-22 CG-1406-51 CG-1406-06 CG-1406-56 CG-1406-09 CG-1406-11 CG-1406-01 CG-1406-27 CG-1406-28 CG-1406-29 CG-1406-23 CG-1406-03 CG-1406-59 CG-1406-35 CG-1406-26 CG-1406-24 CG-1406-05 CG-1406-62 CG-1406-57 CG-1406-31 CG-1406-08 CG-1406-60
80068-408EA 393.66 USD InStock
80068-408 80068-446 80068-402 80068-424 80068-444 80068-422 80068-400 80068-388 80068-428 80068-406 80068-448 80068-404 80068-426 80068-394 80068-470 80068-392 80068-490 80068-398 80068-452 80068-474 80068-430 80068-396 80068-450 80068-472 80068-390 80068-418 80068-434 80068-456 80068-412 80068-478 80068-410 80068-476 80068-432 80068-454 80068-438 80068-416 80068-436 80068-458 80068-414 80068-460 80068-480 80068-442 80068-420 80068-386 80068-440 80068-462 80068-484
Buchner Funnels, Inner Joint, Chemglass
Funnels Filtration Funnels Büchner Funnels
Buchner-style filter funnel with a lower [ST] inner joint and serrated tubulation for connection to a vacuum source.
  • These buchner funnels are made of glass
  • Lower vaccum assembly
  • Based on funnel porosity fine, medium, coarse and extra coarse fritted glasswares are available
Application of these filter funnels varies with the different frit types that is fine and medium fritted glassware used in the application of filtration and extraction while coarse and extra coarse fritted involved in gas dispersion, gas washing and aborption and coarse filtration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty