Would you like to log in again?
Evaporating Flasks, Heavy Wall, Chemglass
Supplier: Chemglass
CG-1512-08 CG-1512-29 CG-1512-10 CG-1512-33 CG-1512-37 CG-1512-40 CG-1512-46 CG-1512-06 CG-1512-60 CG-1512-04 CG-1512-09 CG-1512-21 CG-1512-39 CG-1512-07 CG-1512-P-35 CG-1512-25 CG-1512-05 CG-1512-42 CG-1512-23 CG-1512-57 CG-1512-P-09 CG-1512-03 CG-1512-52 CG-1512-P-05 CG-1512-35 CG-1512-01 CG-1512-02 CG-1512-59 CG-1512-58 CG-1512-55 CG-1512-P-01 CG-1512-56 CG-1512-P-03 CG-1512-54 CG-1512-27 CG-1512-31 CG-1512-51 CG-1512-44 CG-1512-P-07 CG-1512-53
80069-030EA 140.32 USD InStock
80069-030 80068-968 80069-032 80069-010 80069-034 80069-012 80068-984 80068-982 80068-988 80068-966 80068-986 80068-964 80068-992 80068-970 80068-990 80069-024 80069-002 80069-004 80069-026 80069-028 80069-006 89028-578 89028-576 80069-008 80069-020 80069-000 80069-022 80068-996 80068-974 80068-972 80068-994 80068-978 80068-976 80068-998 80068-980 89028-580 80069-036 80069-014 80069-016 80069-018
Evaporating Flasks, Heavy Wall, Chemglass
Flasks Rotary Evaporator Flasks
Evaporating flask for use with rotary evaporators
Flasks have a heavy wall for vacuum applications and are balanced to prevent run-out during rotation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty