Would you like to log in again?
Erlenmeyer Flasks, with Screw Caps, Chemglass
Supplier: Chemglass
CG-1543-04 CG-1543-01 CG-1543-03 CG-1543-05 CG-1543-07 CG-1543-02 CG-1543-08 CG-1543-06
80070-072EA 55.65 USD InStock
80070-072 80070-082 80070-070 80070-080 80070-078 80070-076 80070-074 80070-084
Erlenmeyer Flasks, with Screw Caps, Chemglass
Flasks Erlenmeyer Flasks
Erlenmeyer flask has screw thread finish and long skirted caps that can be loosened without falling off while shaking flask.
Ordering information: Supplied complete with white rubber lined black phenolic cap.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty