Would you like to log in again?
Flowmeters, Bubble, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1607-01 CG-1607-02
80070-428EA 479.87 USD InStock
80070-428 80070-426
Flowmeters, Bubble, Chemglass
Flow Meters
Flowmeter used for the control of carrier gasses.
  • Projection at top breaks bubbles
Hose connection has a outer diameter of 9 mm at the middle ring.
Delivery information: Supplied complete with rubber bulb.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty