Would you like to log in again?
Freeze Dry Flasks, Chemglass
Supplier: Chemglass
CG-1611-09 CG-1611-25 CG-1611-06 CG-1611-24 CG-1611-05 CG-1611-26 CG-1611-22 CG-1611-23 CG-1611-04 CG-1611-08 CG-1611-07 CG-1611-01
80070-438EA 259 USD InStock
80070-438 80070-458 80070-436 80070-446 80070-456 80070-444 80070-454 80070-442 80070-452 80070-430 80070-440 80070-460
Freeze Dry Flasks, Chemglass
Flasks Flat Bottom Flasks
These flat bottom borosilicate flask can be used with Labconco and Virtis freeze dryers.
Wide mouth design provides easy access sample.
Ordering information: Flasks only, not suppled with rubber top, filter or O-ring.
Caution: Labconco and Virtis are not related to or affiliated with Chemglass, and these manufacturers have not approved or validated these products for use on their equipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty