Would you like to log in again?
Addition Funnels with Glass Stopcock, Chemglass
Supplier: Chemglass
CG-1700-03 CG-1700-10 CG-1700-04 CG-1700-01 CG-1700-02 CG-1700-14 CG-1700-16 CG-1700-12 CG-1700-06
80070-478EA 395.8 USD InStock
80070-478 80070-488 80070-476 80070-500 80070-474 80070-496 80070-484 80070-492 80070-480
Addition Funnels with Glass Stopcock, Chemglass
Funnels Dropping Funnels
Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size.
  • With pressure equalizing arm and glass stopcock
Delivery information: Funnels with 14/20 and 19/22 joints are supplied complete with a stopper.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty