Would you like to log in again?
Funnels, Addition, PTFE Stopcocks, Chemglass
Supplier: Chemglass
CG-1702-10 CG-1702-11 CG-1702-08 CG-1702-07 CG-1702-15 CG-1702-05 CG-1702-09 CG-1702-16 CG-1702-17 CG-1702-01 CG-1702-14 CG-1702-12 CG-1702-13 CG-1702-06
80070-530EA 306.06 USD InStock
80070-530 80070-520 80070-532 80070-522 80070-512 80070-534 80070-524 80070-536 80070-514 80070-504 80070-526 80070-516 80070-528 80070-518
Funnels, Addition, PTFE Stopcocks, Chemglass
Funnels Dropping Funnels
Funnels feature a standard taper joint.
  • Standard taper top outer joint and lower inner joint, both being the same size
  • With pressure equalizing arm and PTFE stopcock
Funnels with 14/20 and 19/22 joints are supplied complete with a stopper.
Ordering information: For replacement stoppers see CG-3000.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty