Would you like to log in again?
Addition Funnels, Cooling Jacket, Glass Stopcock, Chemglass
Supplier: Chemglass
CG-1703-07 CG-1703-05 CG-1703-01 CG-1703-04 CG-1703-03 CG-1703-02
80070-550EA 1241.34 USD InStock
80070-550 80070-540 80070-542 80070-544 80070-546 80070-538
Addition Funnels, Cooling Jacket, Glass Stopcock, Chemglass
Funnels Dropping Funnels
Jacket permits the cooling of funnel contents by dry ice. Drain outlet has an 8 mm outer diameter. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty