Would you like to log in again?
Solvent Addition Funnels, Chemglass
Supplier: Chemglass
CG-1758-04 CG-1758-06 CG-1758-02 CG-1758-01 CG-1758-03
80071-192EA 166.22 USD InStock
80071-192 80071-202 80071-198 80071-196 80071-194
Solvent Addition Funnels, Chemglass
Funnels Dropping Funnels
Funnel design permits addition of solvents to flash chromatography columns without disturbing the silica-gel/packing layer.
  • Lower end of funnel is closed
  • Stem contains a series of holes through which the solvent drains
  • Top of funnel is 75 mm (3")
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty