Would you like to log in again?
NMR Tube Cleaner, Morris, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1850-02 CG-1850-09 CG-1850-10 CG-1850-06 CG-1850-08 CG-1850-07 CG-1850-03 CG-1850-01 CG-1850-05 CG-1850-04
80071-328EA 492.92 USD InStock
80071-328 80071-326 80071-338 80071-324 80071-336 80071-322 80071-334 80071-332 80071-340 80071-330
NMR Tube Cleaner, Morris, Chemglass
Tubes NMR Tubes
The Morris NMR tube cleaner is a durable apparatus which will clean 5 mm NMR tubes with as little as 10 ml of solvent in one easy operation.
  • Does not require a stopper on the NMR tube
  • No fragile side arm
Ordering information: Complete item consists of NMR tube washer, adapter, 250 ml HW filter flask and instructions for use.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty