Would you like to log in again?
Peptide Synthesis Vessels, Solid Phase, T-Bore PTFE Stopcocks, Vacuum, Chemglass
Supplier: Chemglass
CG-1866-02 CG-1866-13 CG-1866-10 CG-1866-03 CG-1866-06 CG-1866-14 CG-1866-05 CG-1866-11 CG-1866-12 CG-1866-15 CG-1866-01 CG-1866-04
80071-390EA 405.89 USD InStock
80071-390 80071-388 80071-400 80071-378 80071-386 80071-398 80071-384 80071-396 80071-382 80071-394 80071-380 80071-392
Peptide Synthesis Vessels, Solid Phase, T-Bore PTFE Stopcocks, Vacuum, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Similar to peptide synthesis vessel, but with the addition of a lower standard taper inner joint.
  • This vessel have addition lower standard taper inner joint allow collection into a vessel having a 24/40 outer joint
  • Available with medium or coarse porosity fritted glass resin support
  • A serrated hose connection is sealed on the inner joint for connection to a vacuum source
The 'T' bore stopcock has an 8 mm O.D. sidearm for connection to nitrogen, while the serrated hose connection has an O.D. of 8 mm at the middle ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty