Would you like to log in again?
Peptide Vessels, Jacketed with Lower Drain Valves, Chemglass
Supplier: Chemglass
CG-1868-02 CG-1868-01 CG-1868-04 CG-1868-03
80071-406EA 2130.97 USD InStock
80071-406 80071-404 80071-402 80071-408
Peptide Vessels, Jacketed with Lower Drain Valves, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Cylindrical, jacketed, peptide synthesis reaction vessel with coarse porosity fritted resin support.
  • The vessel is constructed with a flange having a groove formed in the sealing surface of the flange
  • Flange is also ground to permit a greased seal with any of the corresponding lids
  • Lower outlet is a CG-511 bottom drain valve
Cooling jacket is equipped with two removable hose connections to reduce the risk of injury while attaching or detaching flexible tubing.
Ordering information: Supplied complete with two polypropylene hose connections and two GL-14 screw caps (for the clamp see CG-141; for the o-ring see CG-147). Lid not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty