Would you like to log in again?
Reaction Vessels, Cylindrical, Unjacketed, Lower Drain Valves, Chemglass
Supplier: Chemglass
CG-1924-03 CG-1924-01 CG-1924-02 CG-1924-05
80071-468EA 1320.1 USD InStock
80071-468 80071-476 80071-472 80071-470
Reaction Vessels, Cylindrical, Unjacketed, Lower Drain Valves, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Reaction vessels feature groove in the ceiling surface.
  • Cylindrical reaction vessel constructed with a flange
  • Having a groove formed in the sealing surface of the flange
The unique design of the Chem-Drain valve incorporates a PTFE plunger that allows the contents of the flask to drain directly through the bore of the plunger. When closed, the tip of the PTFE plug seals into the lower radius of the flask avoiding the dead space normally associated with standard stopcock drains. This feature permits total mixing and drainage of the contents of the flask.
Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. For clamp see CG-141 or CG-141-T. For O-ring see CG-147.
Ordering information: 500 ml through 4 l sizes are available with morton indents on a special order basis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty