Would you like to log in again?
Spatulas, Rounded, Micro, Non-Magnetic, Chemglass
Supplier: Chemglass
CG-1981-12 CG-1981-10
80071-666EA 15.17 USD InStock
80071-666 80071-664
Spatulas, Rounded, Micro, Non-Magnetic, Chemglass
Spatulas
Spatulas are made of non-magnetic stainless steel and feature a mirror finish.
  • Spatula will pass through a 14/10 or 14/20 outer joint
  • Both ends are rounded and are approximately ⁵/₁₆" (8 mm) wide × 2" (51 mm) in length
  • Non-magnetic
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty