Would you like to log in again?
STIRRER ASMBY. MACRO 445MM
Supplier: Chemglass
STIRRER ASSEMBLY, MACRO, CHEM-STIR^ Joint size: 24/40. For Flask Capacity: 500 to 5000ml. Blade Size: 19x75mm; Shaft Length: 445mm. Assembly consists of a 10mm O.D. Chem-Stir stirrer shaft, a precision bore bearing having a standard taper inner joint and a fluoropolymer resin blade.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STIRRER ASMBY. MACRO 445MM 80071-712 CG-2068-02 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty