Would you like to log in again?
Stirrer Bearings, Macro, Glass, 10 mm, Chemglass
Supplier: Chemglass
CG-2070-01 CG-2070-02 CG-2070-05
80071-724EA 137.58 USD InStock
80071-724 80071-732 80071-726
Stirrer Bearings, Macro, Glass, 10 mm, Chemglass
Stirrers Stirring Tools
Precision bore bearing for use in moderate vacuum with a CG-2074 or CG-2075 ground 10 mm stirrer shaft.
Caution: NOTE: Not for use with CG-2078 or CG-2079 polished stirrer shafts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty