Would you like to log in again?
STIRRER BEARING PTFE 45/50
Supplier: Chemglass
STIRRER BEARING, FLUOROPOLYMER RESIN, CHEM-STIR, 10MM^ Complete Bearing. Joint size: 45/50. New design grease-free fluoropolymer resin stirrer bearing designed for use under moderate vacuum.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STIRRER BEARING PTFE 45/50 80071-770 CG-2076-05 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty