Would you like to log in again?
ADAPTER GLASS ONLY 29/42
Supplier: Chemglass
STIRRER BEARING, FLUOROPOLYMER RESIN, CHEM-STIR, 10MM^ Replacement Components. 29/42 Glass Adapter Only. New design grease-free fluoropolymer resin stirrer bearing designed for use under moderate vacuum.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER GLASS ONLY 29/42 80071-778 CG-2076-12 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty