Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Stirrer Bearings, 10 mm, PTFE, Chemglass
Supplier: Chemglass
CG-2077-04 CG-2077-02 CG-2077-11 CG-2077-10 CG-2077-05 CG-2077-12 CG-2077-13 CG-2077-03 CG-2077-14 CG-2077-01
80071-812EA 121.81 USD InStock
80071-812 80071-802 80071-810 80071-800 80071-798 80071-796 80071-794 80071-808 80071-806 80071-814
Stirrer Bearings, 10 mm, PTFE, Chemglass
Stirrers Stirring Shafts
Grease-Free PTFE stirrer bearing designed for use in moderate vacuum applications
Bearing consists of a threaded glass adapter having the listed inner joint, a saddle o-ring, a body bushing with a compression and locking nut, and a precision PTFE inner bearing.
The saddle o-ring provides only glass and PTFE contact with the reaction, while allowing for the vacuum tight seal. Design of the bearing permits replacement of the saddle o-ring at a fraction of the cost of the entire bearing.
Ordering information: For use only with CG-2078 or CG-2079 Precision Ground and Polished Stirrer Shafts.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty