Would you like to log in again?
ADAPTER GLASS ONLY 24/40
Supplier: Chemglass
STIRRER BEARING, WITH INLET/OUTLET^ Replacement Components. Glass Adapter Only, 24/40. fluoropolymer resin stirrer bearing with a hose connection for vacuum or inert gas source on a single neck flask.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER GLASS ONLY 24/40 80071-824 CG-2077-A-11 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty