Would you like to log in again?
STIRRER BEARING 19MM 45/50
Supplier: Chemglass
STIRRER BEARING, HEAVY DUTY, 19MM^ Joint size: 45/50. Heavy duty 19mm bearing for use in stirring large volumes under high torque. Bearing has a tooled lubricant cup. NOTE: Not for use with CG-CG-2087 or CG-2088 polished shafts.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STIRRER BEARING 19MM 45/50 80071-848 CG-2082-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty