Would you like to log in again?
LOWER FLASK ONLY
Supplier: Chemglass
SUBLIMER, MICRO^ Replacement Components. Lower Flask Only. Micro scale sublimer for use with quantities as small as 0.5 grams. The sublimer has a capacity of 3ml between the lower portion of the cold finger and the interior bottom of the flask.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LOWER FLASK ONLY 80072-110 CG-3030-02 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty