Would you like to log in again?
CONDENSER ONLY 40ML SUBLIMER
Supplier: Chemglass
SUBLIMATOR, VACUUM, DAILEY ^ Replacement Components. Condenser Only, 40ml sublimer. A grease free, two piece sublimation apparatus consisting of an upper water cooled condenser and lower vessel to hold the material to be sublimed.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CONDENSER ONLY 40ML SUBLIMER 80072-134 CG-3034-06 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty