Would you like to log in again?
CONDENSER ONLY 15ML SUBLIMER
Supplier: Chemglass
SUBLIMATION APPARATUS^ Replacement Components. Condenser Only, 15ml Sublimer. Two piece sublimer consisting of a upper water cooled condenser and lower flat bottom flask. Sublimate is collected on the inner wall of the condenser.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CONDENSER ONLY 15ML SUBLIMER 80072-142 CG-3036-04 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty