Would you like to log in again?
VAC. MAN. L 3 PORT 0-8MM VA
Supplier: Chemglass
VACUUM MANIFOLD^ With Left Hand Vacuum Connection. Number of Ports: 3; Valve Size: 0-8mm; Overall Width: 350mm. Single bank general purpose manifold with 3, 4, or 5 ports. Valves are either 0-4 or 0-8mm with CHEM-CAP plug allowing only glass and fluoropolymer resin contact with material being used.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VAC. MAN. L 3 PORT 0-8MM VA 80072-452 CG-4434-08 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty