Would you like to log in again?
Vacuum Trap with Serrated Connections, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-4515-20 CG-4515-03 CG-4515-02 CG-4515-21 CG-4515-01 CG-4515-04
80072-578EA 486.09 USD InStock
80072-578 80072-548 80072-552 80072-554 80072-550 80072-580
Vacuum Trap with Serrated Connections, Chemglass
Vacuum Traps
Two-piece construction for easy removal of condensate
Inlet and outlet are 180° apart for installation into a variety of vacuum systems. Stopper has a [ST] inner joint and body has an outer joint. Body length is measured below the joint. Serrated hose connections have an O.D. of 13mm at the smallest serration and 16mm at the largest serration.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty