Would you like to log in again?
VACUUM TRAP DUAL CHAMBER
Supplier: Chemglass
VACUUM TRAP, DUAL CHAMBER^ Dual Chamber Trap, Complete. Dual chamber vacuum trap affords increased condensing capacity in a much shorter length. The dry ice cold fingers have 55/50 inner joints that fit into the outer joints on the main trap body and have a capacity of approx. 300ml each.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VACUUM TRAP DUAL CHAMBER 80072-630 CG-4525-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty