Would you like to log in again?
VACUUM TRAP BODY 500ML
Supplier: Chemglass
VACUUM TRAP^ Replacement Components. 500ml Trap Body Only. Similar to CG-4529, but having a 40mm o-ring joint to connect the trap body to the stopper. O-ring joint provides for a vacuum-tight grease-free seal and permits the trap body to be easily disconnected from the stopper for cleaning.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VACUUM TRAP BODY 500ML 80072-666 CG-4531-03 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty